This item is now discontinued. Please browse similar items
Fluke Calibration 1653471 Fast Response PRT Probe, 10" , 10" , Bare Wire
Representative image only.
Fluke Calibration Mfr # 5622-10-B – Item # RK-90450-57
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)662
  • Min Temperature (° C)-200
  • Max Temperature (° C)350
  • Temperature Accuracy±0.072°F (±0.04°C)
  • Connector TypeStripped Wire
  • Sheath Diameter (in)0.004
  • DescriptionFast Response PRT Probe, 10\" , 10\" , Bare Wire
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5622 Series - Fast response PRT/RTD probes
Designed for special temperature measurement applications requiring fast response or short immersion over a wide temperature range. These high-quality wire-wound sensors are small allowing heat to transfer to the sensors quickly, with a time constants as fast as 0.4 seconds. Immersion requirements for these probes is also a plus, ranging from just 10 mm to 64 mm (0.4 to 2.5 inch), depending on the model.
Available for sale in the United States and Canada only.