This item is now discontinued. Please browse similar items
Fluke Calibration 1652561 (Scientific) Thermistor Probe, 9" x 1/8" , Bare Wire
Representative image only.
Fluke Calibration Mfr # 5610-9-B – Item # RK-90451-07
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeThermistor
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature Accuracy±0.018°F (±0.01°C)
  • Connector TypeStripped Wire
  • Sheath Diameter (in)0.125
  • Description(Scientific) Thermistor Probe, 9\" x 1/8\" , Bare Wire
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5610 Series - Secondary thermistor probes: The small drift and low calibration uncertainty of only ±0.01°C makes this rugged stainless steel thermistor a wise choice when temperatures of interest are between 0 and 100°C. These probes are ideal for measuring temperature in air, liquid or soil.
Available for sale in the United States and Canada only.