This item is now discontinued. Please browse similar items

Fluke 830 Laser Shaft Alignment Tool 65700-31

Representative image only.
Fluke Mfr # FLUKE-830 – Item # RK-65700-31
Identifies machine misalignment — eliminating costly repairs and downtime
  • Single laser measurement technology ensures fewer errors from backlash and greater data accuracy
  • Compass measurement mode provides flexible, reliable, and repeatable measurements with included activated electronic inclinometer
  • Dynamic machine tolerance check function provides continuous alignment evaluation of adjustments ensuring all are within machine tolerances
  • Auto save and resume functions ensure data protection and quick access for additional flexibility
  • Intuitive user interface makes machine alignments quick and easy
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • External InterfaceUSB host and USB device (slave); RS232 (serial) for sensor; AC adapter/charger socket; Integrated wireless communication, Class 1, transmitting power 100 mW
  • Memory64 MB RAM, 64 MB flash
  • DisplayBacklit LED, 3.5\", 320 x 240 pixel
  • Battery TypeIntegrated Lithium-ion polymer rechargeable battery, 7.4 V/ 2.6 Ah
  • Height (in)6 1/2
  • Width (in)8 3/4
  • Depth (in)1 13/16
  • Height (cm)16.5
  • Width (cm)22.2
  • Depth (cm)4.6
  • DescriptionLaser Shaft Alignment Tool 65700-31
  • Warranty2 Years
Two year warrantyIP 65

Key Features

  • Single laser measurement technology ensures fewer errors from backlash and greater data accuracy
  • Compass measurement mode provides flexible, reliable, and repeatable measurements with included activated electronic inclinometer
  • Dynamic machine tolerance check function provides continuous alignment evaluation of adjustments ensuring all are within machine tolerances
  • Auto save and resume functions ensure data protection and quick access for additional flexibility
  • Intuitive user interface makes machine alignments quick and easy
  • 2 year warranty

More About this Item

The Fluke 830 laser shaft alignment tool provides quick, easy-to-interpret, accurate machine alignment data, with actionable answers, allowing you to keep your plant up and running. Ensuring your rotating machines are in alignment will help reduce energy consumption, seal, coupling, and bearing failures, lower machine vibration levels, and help eliminate shaft cracking and loose foundation bolts.

In contrast to using the straight-edge method or dial indicators, the Fluke 830 tool performs the complicated alignment calculations for you, ensuring you have the answers you need to quickly align your machine. The enhanced user interface provides easy-to-understand results that don’t require extensive alignment knowledge, and provides a unique “All-in-One” result screen that displays both coupling results and feet corrections (vertical and horizontal) in real terms making it easy to take corrective actions.

This instrument allows you to quickly evaluate machine alignment in three simple steps. First, the step-by-step input of your machines dimensions. Second, as you rotate the shaft the instrument receives instant measurement verification. Third, the actual correction numbers are displayed with a four-color severity scale that indicates the degree in which your machine is misaligned. Use the precision shims to save time and perform precision alignments reducing the need for additional diagnostics. Additional features include “clock mode” for vertical and flanged machines, “machine flip” to flip image of machine on screen for alignment jobs with obstructions, and “resume mode” that allows you to save your work and return back at the same step at anytime.

Available for sale in the United States and Canada only

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat