This item is now discontinued. Please browse similar items
Fluke 718EX 30 Intrinsically Safe Pressure Calibrators, -12 to 200 PSI
Representative image only.
Fluke Mfr # FLUKE-718EX 30 – Item # RK-68450-92
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionIntrinsically Safe Pressure Calibrators, -12 to 200 PSI
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Pressure sourcea nd milliamp measurement capability to calibrate and maintain almost any pressure device with one tool
Integrated pump with media protection helps protect the pump from fluid contamination and allows pumps to be serviced in the field
Error % calculation for quicker pass / fail decision making in the field
Best in class mA accuracy (0.015%) for better measurement confidence and workload coverage