This item is now discontinued. Please browse similar items
Fluke 709H/TRACK Loop Calibrator Software
Representative image only.
Fluke Mfr # FLUKE-709H/TRACK – Item # RK-26072-51
Easily upload data from your Fluke 709H calibrator
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLoop Calibrator Software
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Once data is recorded by the 709H meter in the field, the 709H/ TRACK software can upload the HART configuration of up to (20) HART devices in your plant and output data in either (.csv) or (.txt) format.
Available for sale in the United States and Canada only