Fluke 700ILF In-line Filter,1 Micron,100 Ps (PROCESS TOOLS)
Representative image only.
Fluke Mfr # FLUKE-700ILF – Item # RK-68120-70
Isolate calibrator from the incidental contract with fluids
  • 1 year warranty
$248.99CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionIn-line Filter,1 Micron,100 Ps (PROCESS TOOLS)
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The Fluke 700ILF In-line Filter is specifically designed to trap fine particles from atmospheric air, compressed air, and instrument air. It is used with the Fluke 713, 717, or 718 Pressure Calibrators. It can also be used to absorb small quantities of liquid oil and water in droplet form.