This item is now discontinued. Please browse similar items

Fluke 434-II Three-phase Energy Analyzer with Flex Current Probes

Representative image only.
Fluke Mfr # FLUKE-434-II – Item # RK-20043-11
Quickly identify power problems, manage, and reduce energy waste
  • Energy loss calculator for pinpoint true system energy losses in dollars
  • Power inverter efficiency allows simultaneous measurement of AC output power and DC input power for power electronics systems using optional DC clamp
  • Troubleshoot real-time - analyzing trends using the cursors and zoom tools
  • Highest safety rating in the industry - 600 V CAT IV / 1000 V CAT III rated for use at the service entrance
  • Automatic trending ensures that every measurement is always automatically recorded, without any set-up
  • System monitors up to ten power quality parameters on one screen according to EN50160 power quality standard
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Min AC Voltage (Voltage)1
  • Max AC Voltage (Voltage)1000
  • Min AC Current (amps)0.5
  • Max AC Current (amps)6000
  • Min DC Voltage (Voltage)1
  • Max DC Voltage (Voltage)1000
  • Min DC Current (amps)0.5
  • Max DC Current (amps)6000
  • Power Factor Range0 to 1
  • HarmonicsHarmonics volts, THD, harmonic amps, K factor amps, harmonic watts, THD watts, K factor watts, interharmonic amps, Vrms, Arms (relative to fundamental or to total RMS)
  • Watts (AC/DC) Range6000 MW (max)
  • MemoryData is stored on SD card (8 GB incuded; 32 GB max)
  • Sample Rate200 kS/s on each channel simultaneously
  • Display127 x 88 mm (6\" diagonal) LCD; 320 x 240 pixels
  • Battery TypeLi-ion battery; 28 Wh (up to 7 hours continuous use)
  • Height (in)13
  • Width (in)12 1/4
  • Depth (in)2 1/2
  • Height (cm)33
  • Width (cm)31.1
  • Depth (cm)63.5
  • DescriptionThree-phase Energy Analyzer with Flex Current Probes
  • Warranty2 Years
Two year warranty

Key Features

  • Energy loss calculator for pinpoint true system energy losses in dollars
  • Power inverter efficiency allows simultaneous measurement of AC output power and DC input power for power electronics systems using optional DC clamp
  • Troubleshoot real-time - analyzing trends using the cursors and zoom tools
  • Highest safety rating in the industry - 600 V CAT IV / 1000 V CAT III rated for use at the service entrance
  • Automatic trending ensures that every measurement is always automatically recorded, without any set-up
  • System monitors up to ten power quality parameters on one screen according to EN50160 power quality standard
  • 2 year warranty

More About this Item

Available for Sale in the United States and Canada only

The new Fluke 430 Series II three-phase power quality analyzers help you locate, predict, prevent, and troubleshoot problems in three-phase and single-phase power distribution systems. Units feature an exclusive energy loss algorithm, Unified Power Measurement, that measures and quantifies energy losses due to harmonics and unbalance issues, allowing you to pinpoint the origin of energy waste within a system. Troubleshooting is faster with on-screen display of trends and captured events, even while background recording continues. The IEC standards for flicker, harmonics and power quality, are all built-in taking the guesswork out of power quality.

All models feature volt, amp, hertz, dips and swells, harmonics, power and energy, energy loss calculator, unbalance, monitor, power inverter efficiency, and inrush capabilities.

What's included: Test lead and alligator clip set (TL430), four 24" (61 cm L) flexible current probes (i430), software, 8 GB SD card, USB cable, soft carrying case, power adapter (BC430), international plug adapter set, and Li-ion battery (BP290).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat