This item is now discontinued. Please browse similar items
Fluke 345 Digital Power Quality Clamp Meter
Representative image only.
Fluke Mfr # FLUKE-345 – Item # RK-20003-38
Quickly troubleshoot power quality issues with the easy-to-use combination of a power quality meter, current clamp, and data logger
  • True RMS AC and DC clamp-on current measurements up to 2000 A (1400 A RMS)
  • Make effective measurements on variable speed drives and other noisy equipment with the help of the switchable low-pass filter
  • Highest safety rating in the industry - 600 V CAT IV / 1000 V CAT III
  • Troubleshoot onscreen with color display of waveforms, harmonics, and recorded data
  • Investigate intermittent issues with logging of min, max, and average measurements
  • Measures V, A, Hz, CF,THD, DF, W, VA, VAR, kWh and Power factor even on distorted waveforms
  • Assess the effects of loads with DC ripple measurement
  • Quantify demand with three-phase power capability for balanced loads
  • Inrush mode for troubleshooting nuisance circuit breaker tripping
  • View graphs and generate reports with included Power Log software
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDigital Power Quality Clamp Meter
  • Warranty2 Years
Two year warranty
Key Features
  • True RMS AC and DC clamp-on current measurements up to 2000 A (1400 A RMS)
  • Make effective measurements on variable speed drives and other noisy equipment with the help of the switchable low-pass filter
  • Highest safety rating in the industry - 600 V CAT IV / 1000 V CAT III
  • Troubleshoot onscreen with color display of waveforms, harmonics, and recorded data
  • Investigate intermittent issues with logging of min, max, and average measurements
  • Measures V, A, Hz, CF,THD, DF, W, VA, VAR, kWh and Power factor even on distorted waveforms
  • Assess the effects of loads with DC ripple measurement
  • Quantify demand with three-phase power capability for balanced loads
  • Inrush mode for troubleshooting nuisance circuit breaker tripping
  • View graphs and generate reports with included Power Log software
  • 2 year warranty
More About this Item
The Fluke 345 Power Quality Clamp Meter is the ideal tool for servicing and commissioning electronic switching loads such as drives and UPS systems. The practical clamp-on design quickly displays the most common power quality parameters. For long-term applications, the Fluke 345 records parameters for viewing on-screen or with included Power Log software.
Sold in the United States and Canada only