Harmonics Accuracy≥1 V: ±5% of reading; >1 V: 0.05 V
Total Harmonic Distortion (Thd)10
DataloggingInternal flash memory (not user replaceable)
MemoryTypical 20 logging sessions of 4 weeks with 1-minute intervals and 500 events
Communication PortsEthernet, USB (mini B), and Wi-Fi
Battery Type1 3.6V Lithium and 1 1/2 AA (included)
Length (in)9 1/8
Height (in)2 1/8
Width (in)7 1/8
Length (cm)23
Height (cm)5.4
Width (cm)18
DescriptionThree-Phase Power Quality Logger, 24\"L iFlex 1500 A probe
Includes
Four 24"L iFlex 1500 A probes, four black dolphin clips, set of wire clips, 3-phase + N voltage test lead, set of two 7" (18 cm) L test leads, set of two 6.6 ft (1.5 m) L test leads, USB cable with mini connector, USB drive with software, two Wi-Fi/BLE-to-USB adapters, cable marker kit, soft storage case, voltage input adapter, and AC power cable.
Key Features
Easy access to power quality and energy data in real time
Wireless transfer of data minimizes your time in potentially hazardous environments
Quickly create standardized reports for analysis
Meets IEC 6100-4-30 Class A Edition 3 standard
Measures basic power quality parameters for analysis
Includes four iFlex 1500 A probes
2 year warranty
More About this Item
Measure all three voltage and current phases plus the neutral current as well as power and frequency with these power quality loggers. Loggers simultaneously measure up to 500 parameters while capturing events to identify intermittent and hard-to-find power quality issues. Power and power quality variables are automatically logged so you don't have to worry about losing data. Order Fluke model 1746 or 1748 to measure key power quality parameters such as harmonics and interharmonics. Advanced Fluke model 1748 also measures waveform deviations.
Download your data in multiple ways. With both Wi-Fi and Ethernet connection capability, you can use it in hard-to-reach areas as well as in areas where Wi-Fi may be spotty. Loggers feature two external USB ports—one for PC connection and another for quick data download to a USB device. Included software lets you download and quickly analyze measured values and perform advanced analysis to help make decisions in real time. Easily create standardized reports according to commonly used standards such as EN 50160 and IEEE 519.
Easily place in tight spaces and panels—loggers only measure 9-1/8" x 7-1/8" x 2-1/8". Rugged loggers retain IP65 rating when used with IP65 voltage input adapter (included). Safely secure loggers with a standard chain or other security device.
Fluke model 1742 three-phase power quality logger measures voltage, current, power, power factor, frequency, energy forward/reverse, peak demand, THD and flicker.
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.