Fluke 1587 Kit/62MAX+ FC Wireless Electrical Troubleshooting Kit
Representative image only.
Fluke Mfr # 1587KIT/62MAX+ FC – Item # RK-20050-88
Save money and reduce costly downtime
  • Kit includes:
  • 1587 FC Insulation/Multimeter
  • i400 Current clamp
  • 62 MAX+ IR thermometer
  • One remote probe, three general probes, one test lead, and three alligator clips
  • General purpose temperature probe with K-type thermocouple
  • Hard carrying case
  • Available for sale in the United States and Canada only
  • 3 year warranty
$1,887.94CAD / Each
$266.68 - $374.24CAD / Each
for an additional
$266.68CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWireless Electrical Troubleshooting Kit
  • Warranty3 Years
3 Year Warranty
Key Features
  • Kit includes:
  • 1587 FC Insulation/Multimeter
  • i400 Current clamp
  • 62 MAX+ IR thermometer
  • One remote probe, three general probes, one test lead, and three alligator clips
  • General purpose temperature probe with K-type thermocouple
  • Hard carrying case
  • Available for sale in the United States and Canada only
  • 3 year warranty
More About this Item
This advanced wireless electrical kit is ideal for work on motors, cables, switch gear and is a vital part of any preventive maintenance tool program. Easily perform insulation tests plus a wide-range of DMM tasks with confidence and ease. The i400 clamp along with the 1587 FC allows you to accurately measure AC current without breaking the circuit. This kit will help you eliminate downtime and keep your equipment running smoothly.