This item is now discontinued. Please browse similar items
Flir 71001-1103 Explosion Proof Automation Thermal Camera, F=18mm and 25 Deg Lens
Representative image only.
Flir Mfr # 71001-1103 – Item # RK-39755-31
Safely monitor critical assets in explosive atmospheres
  • Ideal for use in harsh, dusty environment-- IP67 rated
  • Heater prevents fogging and freezing of protection window
  • Plug and play installation
  • 320 x 240 pixel IR resolution
  • MPEG-4 streamed video
  • C-Video
  • Extensive analysis functions
  • Built in alarm functions
  • Ethernet/IP connectivity
  • Digital inputs/outputs
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Pixels / Thermal Resolution320 x 240
  • Field Of View (FOV)25 degrees
  • Min Temperature (° F)-4
  • Max Temperature (° F)662
  • Min Temperature (° C)-20
  • Max Temperature (° C)350
  • Zoom1 to 8× continuous, digital, interpolating zooming on images
  • Thermal Sensitivity (N.E.T.D) (° F)≤ 0.09 at 86
  • Thermal Sensitivity (N.E.T.D) (° C)≤ 0.05 at 30
  • Focus TypeAutomatic or manual
  • Min Focus Distance (m)0.4
  • Camera Included A310ex Explosion Proof Automation Thermal Camera, f=18mm and 25 deg Lens
  • DescriptionExplosion Proof Automation Thermal Camera, F=18mm and 25 Deg Lens
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 67
Key Features
  • Ideal for use in harsh, dusty environment-- IP67 rated
  • Heater prevents fogging and freezing of protection window
  • Plug and play installation
  • 320 x 240 pixel IR resolution
  • MPEG-4 streamed video
  • C-Video
  • Extensive analysis functions
  • Built in alarm functions
  • Ethernet/IP connectivity
  • Digital inputs/outputs
  • 1 year warranty
More About this Item
FLIR A310 ex is an ATEX compliant solution, with a thermal imaging camera mounted in an enclosure, making it possible to monitor critical and other valuable assets also in explosive atmospheres. Process monitoring, quality control and fire detection in potentially explosive locations are typical applications for the A310 ex. Explosive atmospheres need to be protected from ignition sources by selecting equipment and protective systems which meet the requirements of the ATEX Product Regulations or similar regulations. Flame-proof enclosure prevents any explosion transmission from the inside of the enclosure to the outside.
The integrated controller features several digital I/O channels and sensors for temperature, humidity and pressure. Among other functions, the I/O channels enable the user to switch on/off the camera and the heater via remote control. The access is accomplished through an integrated web interface or Modbus TCP/IP. The controller is equipped with two fiber optic and two Ethernet parts. This enables a flexible network integration in star or ring topologies.
Automation thermal imagers in many cases require application support prior to ordering. Please contact our technical application group or sales team for support on this product. In some areas of the country onsite field assessments can be arranged.
he FLIR A310 camera offers an affordable and accurate temperature measurement solution for anyone who needs to solve problems that do not call for the highest speed or reaction and who uses a PC. Due to to its composite video output, it is also an excellent choice for thermal image automation applications, where you can utilize its unique properties such as looking through steam.
Package Contents:
• Infrared camera with lens
• Calibration certificate
• Cardboard box
• Downloads brochure
• Ethernet cable
• FLIR apps card
• FLIR Tools download card
• Getting Started Guide
• Important Information Guide
• Mains cable
• Optics brochure
• Power cable, pig-tailed
• Power supply
• Service and training brochure
• Thank you card
• User documentation CD-ROM
• Utility CD-ROM
• Registration card
Available for sale in the United States and Canada only