This item is now discontinued. Please browse similar items
Flir A310 PT Pan and Tilt Automation Thermal Camera with 6° Lens
Representative image only.
Flir Mfr # FLIR A310PT W/6 LENS – Item # RK-39755-63
Detect potential threats before they happen
  • IP-66 rated housing
  • Day/night 36 x zoom color CCD
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Detector Size340 x 240
  • Field Of View (FOV)6° x 4.5°
  • Min Temperature (° F)-4
  • Max Temperature (° F)662
  • Min Temperature (° C)-20
  • Max Temperature (° C)350
  • Thermal Sensitivity (N.E.T.D) (° F)≤ 0.09 at 86
  • Thermal Sensitivity (N.E.T.D) (° C)≤ 0.05 at 30
  • Camera Included A310pt 7 Pan and Tilt with 6 deg Lens
  • DescriptionPan and Tilt Automation Thermal Camera with 6 Deg Lens
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 66 Rated
Key Features
  • IP-66 rated housing
  • Day/night 36 x zoom color CCD
  • 1 year warranty
More About this Item
The A310 PT dual-sensor cameras combine thermal and visible light video security imaging with automated safety monitoring based on the detection of temperature measurement thresholds. By measuring slight changes in surface temperature, the cameras can detect hotspots or overheating equipment. An alarm alerts you if potential threats exceed pre-set levels.
Due to its composite video output, it is also an excellent choice for thermal image automation applications, where you can utilize its unique properties such as looking through steam.
Available for sale in the United States and Canada only