This item is now discontinued. Please browse similar items
Fisher WLT400 Water Level Indicator 400FT With 1/2inDIA Brass Probe
Representative image only.
Fisher Mfr # WLT400 – Item # RK-99050-18
Contact of the electrode assembly with the water surface is indicated by an LED and the sound of the audio alert beeper
  • Depth indication is provided by numbered metal tags securely crimped to the cable at intervals of five feet
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Depth (ft)400
  • DescriptionWater Level Indicator 400FT With 1/2inDIA Brass Probe
  • Warranty2 year
Two year warranty
Key Features
  • Depth indication is provided by numbered metal tags securely crimped to the cable at intervals of five feet
  • 2 year warranty
More About this Item
The brass electrode assembly is 3/8” in diameter and is easily replaceable in the field. The cable is two-conductor, steel and copper plated, with a tensile strength of approximately 300 lbs and a tough plastic type insulating jacket. It is mounted on a small hand-held spool and stand which can be used conveniently to let out or withdraw the cable and electrode assembly.