This item is now discontinued. Please browse similar items
Federal Signal LP4-09-028-CP Low-profile Audible Alarms, Mini Alarm
Representative image only.
Federal Signal Mfr # LP4-09-028-CP – Item # RK-10404-50
Compact, multi-tone alarms for any application
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)3
  • Diameter (in)3 43/64
  • Height (cm)7.62
  • Diameter (cm)9.32
  • DescriptionLow-profile Audible Alarms, Mini Alarm
IP54 Rated
More About this Item
Audible indication is a useful alarm alternative that is often overlooked. An audible indication alerts the environment to important changes in conditions including fluid level, pH, pressure, temperature, and many others. Low-profile audible alarms provide an audible tone or siren when a change in condition is detected. Choose from a selection of tones and sirens within each unit. Units are mountable to panel or wall. Often people may use visual clues to notate a change in a process or procedure. But adding audible indication, or alarm, can help lab professionals detect improvements or changes faster and with more reliability. Audible indication can be something as simple as a bell, or whistle. For visual alerts try low-profile incandescent lights. These feature a steady burning incandescent light and can be surface or panel mounted. Indicators provide visual alert to a change in condition. Multiple colors are available. For more information about adding an audible indication to your system contact a Davis Application Specialist today.
Mini Alarm 10404-50 has 28 tones—switch selectable during installation with volume control for 20 dB of adjustment. The NEMA 4X, IP54 enclosure with rugged ABS casing and encapsulated electronics makes alarms suitable for outdoor use. Ideal for most PLC applications—excellent for monitoring fluid levels, pH, pressure, temperature, or other systems. Operates in temperatures from -40 to 176°F (-40 to 80°C).