This item is now discontinued. We recommend the following as suggested replacements: RK-20005-34
Extech TK430 True RMS Multimeter and Clamp Meter Electrical Test Kit
Representative image only.
Extech Mfr # TK430 – Item # RK-20005-40
Save time and money with this multifunctional kit!
  • Low current capability—measure down to 0.1 µA
  • True RMS current/voltage
  • Audible and visual input alerts
  • RELATIVE function for establishing a base line reference
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Safety RatingCAT III 600V, CAT III 1000V, EN 61010-1
  • DescriptionTrue RMS Multimeter and Clamp Meter Electrical Test Kit
  • Warranty1 Year
1 Year WarrantyUL Listed.
Key Features
  • Low current capability—measure down to 0.1 µA
  • True RMS current/voltage
  • Audible and visual input alerts
  • RELATIVE function for establishing a base line reference
  • 1 year warranty
More About this Item
Kit includes:
EX430 Autoranging True RMS multimeter
MA200 400 A Clamp meter with basic AC Current and 1 mA resolution
40130 Noncontact voltage detector (detects 100 VAC to 600 VAC)
Set of CAT III-1000 V test leads
Type K thermocouple bead probe and Type K to banana input adaptor
Storage case