This item is now discontinued. Please browse similar items
Extech LED Light Tip Replacement for RD300 Refrigerant Leak Detector
Representative image only.
Extech Mfr # RD300-L – Item # RK-10112-27
See what you’re doing with handy LED light located right on probe tip
  • Color-coded lights visually indicate leak size
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLED Light Tip Replacement for RD300 Refrigerant Leak Detector
  • Warranty6 month
6 Month Warranty
Key Features
  • Color-coded lights visually indicate leak size
  • 6 month warranty
More About this Item
Detect refrigerant leaks in air conditioning and cooling systems with the Extech RD300 leak detector. A convenient LED work light at the tip of the probe illuminates dimly lit areas, making it easier to identify the source of a leak. Select from three sensitivity levels with multicolored light bars visually indicating the leak size. Mute button toggles the audible alarm on and off. Detector also features a flexible probe, low-battery indication, and auto-shutoff.