This item is now discontinued. Please browse similar items
Extech HDV-TX2 6 mm Articulating VideoScope Camera Head with 2m Semi-Rigid Cable
Representative image only.
Extech Mfr # HDV-TX2 – Item # RK-68517-27
Narrow-diameter cameras deliver high image resolution in hard-to-reach areas
  • Compatible with Extech videoscopes and borescopes
  • 2m semi-rigid nondetachable probe (camera head)
  • 6mm diameter camera, 60° FOV, Macro
  • Articulation knob
  • Connects to HDV600 VideoScope with patch cable (HDV-PC)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description6 mm Articulating VideoScope Camera Head with 2m Semi-Rigid Cable
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Compatible with Extech videoscopes and borescopes
  • 2m semi-rigid nondetachable probe (camera head)
  • 6mm diameter camera, 60° FOV, Macro
  • Articulation knob
  • Connects to HDV600 VideoScope with patch cable (HDV-PC)
  • 1 year warranty
More About this Item
Enhance the versitility of your inspection camera with these optional camera heads available with longer cables.