This item is now discontinued. Please browse similar items
Extech EX540 True RMS Industrial Multimeter/Datalogger
Representative image only.
Extech Mfr # EX540 – Item # RK-20034-48
  • 1000V input protection on all functions
  • Dual sensitivity frequency functions
  • Large backlit LCD with bar graph
  • Diode open circuit voltage of 2.8V DC
  • Auto power off with disable feature
  • Min/Max/Relative/Data Hold
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Max (Volts)2.8
  • AC Max (Volts)2.8
  • Safety RatingCAT IV 600V, EN 61010-1
  • DescriptionTrue RMS Industrial Multimeter/Datalogger
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneIP 67
Key Features
  • 1000V input protection on all functions
  • Dual sensitivity frequency functions
  • Large backlit LCD with bar graph
  • Diode open circuit voltage of 2.8V DC
  • Auto power off with disable feature
  • Min/Max/Relative/Data Hold
  • 1 year warranty
More About this Item
EX540 Additional Features include:
Data acquisition mode for real time data transmission to PC