This item is now discontinued. Please browse similar items
Extech ETK35 Electrical Test Kit with AC/DC Clamp Meter
Representative image only.
Extech Mfr # ETK35 – Item # RK-20046-49
Convenient test kit saves you time and money
  • True RMS autoranging multimeter measures AC/DC voltage (600 V), AC/DC current (10 A with resolution to 0.1 µA), and resistance
  • True RMS 400 A AC/DC clamp meter includes built-in noncontact voltage detector with LED alert
  • Clamp meter jaw size of 1.2” (30 mm) for conductors up to 350 MCM
  • Type K temperature bead wire probe for added flexibility
  • Both meters feature continuity/diode test, data hold, max hold, and auto power off
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionElectrical Test Kit with AC/DC Clamp Meter
  • Warranty1 Year
1 Year Warranty
Key Features
  • True RMS autoranging multimeter measures AC/DC voltage (600 V), AC/DC current (10 A with resolution to 0.1 µA), and resistance
  • True RMS 400 A AC/DC clamp meter includes built-in noncontact voltage detector with LED alert
  • Clamp meter jaw size of 1.2” (30 mm) for conductors up to 350 MCM
  • Type K temperature bead wire probe for added flexibility
  • Both meters feature continuity/diode test, data hold, max hold, and auto power off