This item is now discontinued. Please browse similar items
Extech DCP42 DC Switching Power Supply, 160W
Representative image only.
Extech Mfr # DCP42 – Item # RK-20046-45
High efficiency electrical power conversion for greater energy conservation
  • Constantly calculates and adjusts the voltage and current limit points according to available maximum power
  • Dual action rotary encoder control with push knob for fine and coarse tuning
  • Remote control allows programming of output voltage/current, and on/off with included software
  • Three on-board user-defined voltage/current presets provide additional flexibility
  • Store 20 presets with up to 999 cycles each on a PC via USB and included software
  • At maximum current (10 A), voltage is limited to 16 V; at maximum voltage (42 V), current is limited to 3.8 A
  • Software includes DC ramp and waveform generator
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Output Voltage42 VDC
  • Output Current10 A
  • Ripple and Noise≤ 5 mVrms, < 50 mVp-p
  • Power Rating (watts)160
  • DescriptionDC Switching Power Supply, 160W
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Constantly calculates and adjusts the voltage and current limit points according to available maximum power
  • Dual action rotary encoder control with push knob for fine and coarse tuning
  • Remote control allows programming of output voltage/current, and on/off with included software
  • Three on-board user-defined voltage/current presets provide additional flexibility
  • Store 20 presets with up to 999 cycles each on a PC via USB and included software
  • At maximum current (10 A), voltage is limited to 16 V; at maximum voltage (42 V), current is limited to 3.8 A
  • Software includes DC ramp and waveform generator