Expo Engineering HDD-55/120V Heated Drum Dolly, Fits 55gal, 550F, 120 V
Representative image only.
Expo Engineering Mfr # HDD-55/120V – Item # RK-09500-25
Mobile drum heater provides operator flexibility
  • Dolly supports up to 900 pound capacity
  • 1 year warranty
PRODUCT OPTIONS
$1,650.20CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Drum MaterialMetal
  • Drum Size55 gallons
  • Max Temperature (° F)550
  • Max Temperature (° C)288
  • Min TemperatureAmbient;Ambient
  • Plug TypeUS Standard
  • Power (VAC)120
  • Power (amps)14.6
  • Power (watts)1750
  • Diameter (in)22.3 in
  • DescriptionHeated Drum DOlly, Fits 55gal, 550F, 120 V
  • Warranty1 year
1 Year Warranty
Key Features
  • Dolly supports up to 900 pound capacity
  • 1 year warranty
More About this Item
Use this drum heated dolly to warm your nonflammable, nonhazardous material contained within 55-gallon metal drums. The built-in infinite control (0 to 100% adjustable) features on/off as well as heat pulsing action. Drums rest on a spring-loaded 15 1/4” square by 1/4” thick aluminum heat platen, and rolls on 3"-diameter steel swivel casters that feature two-wheel locking capability. Measure the temperature of drum contents with an external temperature measurement device (not included) for more precise temperature control.