This item is now discontinued. Please browse similar items
Eurotherm IQRL-2004 Rail AC Power Distribution Bus; 4 Modules
Representative image only.
Eurotherm-Action Instruments Mfr # IQRL-2004 – Item # RK-68479-05
Provides Power to Action I/Q Signal Conditioning Modules
  • Reduces Wiring Time
  • Improves Reliability with Fewer Wire to Terminal Connections
  • Attaches Directly to the ActionI/Q Module
  • SnapLoc Plug-In Terminals
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Input typeIsolation: 1800V DC, between power conductors
  • Power Supply (VAC)120/240
  • Power Supply (VDC)24
  • ConnectionSocketed screw terminals for 14-22 AWG
  • Max Operating Temperature (° C)65
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° F)131
  • Min Operating Temperature (° F)32
  • DescriptionIQRL-2004 Rail AC Power Distribution Bus; 4 modules
  • Warranty3 month
3 Month Warranty
Key Features
  • Reduces Wiring Time
  • Improves Reliability with Fewer Wire to Terminal Connections
  • Attaches Directly to the ActionI/Q Module
  • SnapLoc Plug-In Terminals
  • 3 month warranty
More About this Item
These are designed to power 4-wire transmitters in the Action I/Q family, specifically models Q1x6, and Q4x6.
The Action I/Q modules are designed to hold the I/QRail firmly in place and parallel to the DIN mounting rail. No additional mounting holes or cutouts are required for I/QRail installation. Action I/Q modules mount on standard TS32 (32mm) or TS35 (35mm) DIN rails which then support the complete installation.
Model 68478-36, IQRL-2002, powers 2 AC modules, and model 68479-05, IQRL-2004, powers 4 AC modules.