This item is now discontinued. Please browse similar items
Eurotherm COMMS BOARDEI-BISY RS422
Representative image only.
Eurotherm Mfr # SUB22/2FE – Item # RK-94470-29
General Purpose PID Temperature Controllers
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The 2200E series are configurable for PID, On/ Off or motorized valve control for heat only or heat/cool applications, satisfying both electrical and gas heating requirements. Self-tuning and customized cooling algorithms are included to optimize control performance. These units employ unique INSTANT ACCURACY™ technology on scaleable universal inputs accepting nine standard thermocouple and RTD inputs, DC process inputs or downloadable custom inputs. A simple ramp and hold profile can be programmed using the internal ramp generator and dwell timer. With custom cooling algorithms, these controllers are ideal for plastics extrusion.