Eppendorf 22-60-002-8 Microcentrifuge Tube, 1.5 mL, Natural Color
Representative image only.
Eppendorf Mfr # 22-60-002-8 – Item # RK-02560-35
$76.62CAD / Pkg of 100
BUY MORE & SAVE
  • 1-5$76.62CAD / Pkg of 100
  • 6-29$68.96CAD / Pkg of 100
  • 30-119$65.13CAD / Pkg of 100
  • 120+$61.30CAD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g25000
  • SterileYes
  • AutoclavableYes
  • Tube Capacity (mL)1.5
  • Tube MaterialPolypropylene
  • Tube ColorNatural
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap MaterialPolypropylene
  • DescriptionMicrocentrifuge Tube, 1.5 mL, Natural Color
More About this Item
Eppendorf® Biopur™ tubes have all the features of the standard and Safe-Lock™ tubes and are individually wrapped and certified sterile. Biopur™ tubes are pyrogen-, RNase-, DNA-and ATP-free; supplied with a quality assurance card.