Eppendorf 022653121 Fixed-angle 4 x 8-tube PCR-strips Rotor, with lid
Representative image only.
Eppendorf Mfr # 022653121 – Item # RK-02580-52
Optional rotors add versitility
PRODUCT OPTIONS
$1,172.82CAD / Each
Often Bought with...
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g18615
  • Rotor TypeFixed-angle
  • Tube Sizes Accepted8-tube PCR strips
  • Number Of Tubes Accepted4
  • DescriptionFixed-angle 4 x 8-tube PCR-strips Rotor, with lid
More About this Item
These optional rotors are compatible with the Eppendorf 5424/5424R variable-speed microcentrifuges. Centrifuge sold separately.
Aerosol-tight rotor is suitable even for hazardous materials, and contoured bowl design minimizes noise.