Eppendorf 022653083 Spin Column Rotor Kit 18 x 1.5/2.0 mL, including lid
Representative image only.
Eppendorf Mfr # 022653083 – Item # RK-02580-50
Optional rotors add versitility
PRODUCT OPTIONS
$1,172.82CAD / Each
Often Bought with...
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g18111
  • Rotor TypeFixed-angle
  • Tube Sizes Accepted1.5 or 2.0 mL microtubes
  • Number Of Tubes Accepted18
  • DescriptionSpin Column Rotor Kit 18 x 1.5/2.0 mL, including lid
More About this Item
These optional rotors are compatible with the Eppendorf 5424/5424R variable-speed microcentrifuges. Centrifuge sold separately.
Aerosol-tight rotor is suitable even for hazardous materials, and contoured bowl design minimizes noise.