Eppendorf 022653041 Fixed-angle 24 x 1.5/2.0 mL Rotor with chemical resistant (PTFE) coating
Representative image only.
Eppendorf Mfr # 022653041 – Item # RK-02580-54
Optional rotors add versitility
PRODUCT OPTIONS
$1,670.82CAD / Each
Often Bought with...
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g21130
  • Rotor TypeFixed-angle
  • Tube Sizes Accepted1.5 or 2.0 mL microtubes
  • Number Of Tubes Accepted24
  • DescriptionFixed-angle 24 x 1.5/2.0 mL Rotor with chemical resistant (PTFE) coating
More About this Item
These optional rotors are compatible with the Eppendorf 5424/5424R variable-speed microcentrifuges. Centrifuge sold separately.
Aerosol-tight rotor is suitable even for hazardous materials, and contoured bowl design minimizes noise.