Eppendorf 022636146 6 x PCR-strip rotor (30 x 1.5 to 2.0 mL tubes)

Representative image only.
Eppendorf Mfr # 022636146 – Item # RK-17483-53
Optional rotors add versitility

PRODUCT OPTIONS

$2,101.05CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Speed (rpm)14000
  • Max RCF x g20800
  • Rotor TypeFixed-angle
  • Tube Sizes Accepted1.5 or 2.0 mL microtubes
  • Number Of Tubes Accepted30
  • Description6 x PCR-strip rotor (30 x 1.5 to 2.0 mL tubes)

More About this Item

These optional rotors are compatible with the Eppendorf 5424/5424R variable-speed microcentrifuges. Centrifuge sold separately.
Aerosol-tight rotor is suitable even for hazardous materials, and contoured bowl design minimizes noise.

Rotors have a safety cover to minimize air turbulence. Rotors and covers are autoclavable. All rotors can be used with optional tube adapters (sold separately).

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat