This item is now discontinued. Please browse similar items
Eppendorf Filter Tips, 0.1-10 µL, medium
Representative image only.
Eppendorf Mfr # 0030078519 – Item # RK-18888-20
Protect your pipettor and prevent cross-contamination
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)10
  • Min Volume (µL)0.1
  • Min Volume (mL)0.0001
  • Max Volume (mL)0.01
  • Tip Volume (mL)0.01
  • Min Dispensing (mL)0.0001
  • Max Dispensing (mL)0.01
  • Color CodeGray
  • Package Of10 racks; 96 tips per rack
  • DescriptionFilterTips,0.1to10µL,medium
More About this Item
Integrated hydrophobic filters do not contain "self-sealing" additives. All filter tips are sterile and PCR clean (free of DNA, DNase, RNase, and PCR inhibitors). Packaged in racks of 96 tips.