Environmental Express Ion Selective Electrode (ISE), Double-Juntion, Glass; Ammonium
Representative image only.
Environmental Express Ion Selective Electrode (ISE), Double-Juntion, Glass; Ammonium
Environmental Express | Item: 3580203
A wide variety of ion selective electrodes are availabe to suit a wide array of applications
$1,119.33CAD / Each
$1,119.33CAD / Each
Buy It Together
Current Item
Environmental Express Ion Selective Electrode (ISE), Double-Juntion, Glass; Ammonium
Environmental Express Ion Selective Electrode (ISE), Double-Juntion, Glass; Ammonium
3580203
$1,119.33CAD / Each
+ + +
Environmental Express EC 2000 Benchtop Meter Kit
3541960
$1,968.45CAD / Each
+ + + + =
Total Price 8339.60 ADD 8 ITEMS TO CART
Product Description
Key Features
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
  • 1 year warranty
More About this Item
Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction, membrane
  • Electrode Body MaterialGlass
  • Ion-Selective ElectrodeAmmonium
  • ConnectorBNC
  • DescriptionIon Selective Electrode (ISE), Double-Juntion, Glass; Ammonium