Environmental Express Air Monitoring PVC Filters, Cleaned, 37 mm Dia; 25/PK
Representative image only.
Environmental Express Item # RK-97016-45
Use for air monitoring of hexavalent chromium
  • NIOSH 7600 OSHA ID-215 Method IOM sampler
  • Made of PVC
  • 37 mm diameter
$197.07CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPVC
  • Diameter (mm)37
  • DescriptionAir Monitoring PVC Filters, Cleaned, 37 mm Dia; 25/PK
Includes
Supplier Certificate of Analysis (CoA) is included with product shipment.
Key Features
  • NIOSH 7600 OSHA ID-215 Method IOM sampler
  • Made of PVC
  • 37 mm diameter
More About this Item
PVC filters are cleaned using a patented process to remove chromium*.
*Licensed from the United States of America under U.S. Patent No. 8,415,454, Rubenstein, M. “Hexavalent chromium and total chromium removal from polyvinylchloride (PVC) polymers”.