This item is now discontinued. Please browse similar items
Endecotts ZSICSA-.710 Sieve Calibration Standard, soda-lime glass, 25 mesh
Representative image only.
Endecotts Mfr # ZSICSA-.710 – Item # RK-59945-07
Spheres are traceable to the United Kingdom National Physical Laboratory
  • Calibrate sieves to an accuracy of ±1µm
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • U.S. Standard Screen Size (No.)25
  • Tyler Standard Screen Size (mesh)24
  • Min Sieve Aperture (microns)600
  • Max Sieve Aperture (microns)850
  • Weight (g)12.5
  • DescriptionSieve Calibration Standard, soda-lime glass, 25 mesh
  • Warranty6 Months
6 Month WarrantyISO9001 Certified Supplier
Key Features
  • Calibrate sieves to an accuracy of ±1µm
  • 6 month warranty
More About this Item
Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture. Standards include a calibration chart.