Endecotts ZSICSA-.063 Sieve Calibration Standard, soda-lime glass, 230 mesh
Representative image only.
Endecotts Mfr # ZSICSA-.063 – Item # RK-59945-21
Spheres are traceable to the United Kingdom National Physical Laboratory
  • Calibrate sieves to an accuracy of ±1µm
  • 6 month warranty
PRODUCT OPTIONS
$462.64CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • U.S. Standard Screen Size (No.)230
  • Tyler Standard Screen Size (mesh)250
  • Min Sieve Aperture (microns)53
  • Max Sieve Aperture (microns)75
  • Weight (g)5
  • DescriptionSieve Calibration Standard, soda-lime glass, 230 mesh
  • Warranty6 Months
6 Month WarrantyISO9001 Certified Supplier
Key Features
  • Calibrate sieves to an accuracy of ±1µm
  • 6 month warranty
More About this Item
Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture. Standards include a calibration chart.