This item is now discontinued. Please browse similar items
Endecotts Sieveware Software for Windows, Single-user License
Representative image only.
Endecotts Mfr # SIEVEWARE – Item # RK-58886-84
Optimize your sieving with a wide range of equipment
  • Wet sieving conversion kits allow you to sieve wet material
  • SieveWare software automatically controls the measurement and weighing procedures
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOctagon 200CL
  • DescriptionSieveWare Software
Key Features
  • Wet sieving conversion kits allow you to sieve wet material
  • SieveWare software automatically controls the measurement and weighing procedures
More About this Item
These accessories ensure that Endecotts sieve shakers can function in a variety of applications.
SieveWare software is perfect for particle size analysis. The Octagon 200CL can be automatically controlled with SieveWare via RS-232 conenction. This software registers, evaluates, and administers measurement data in accordance with different protocol standards. The data can then be transformed into a variety of charts and tables. Data can be printed, saved, or exported.