EMD Millipore UFC901024 Amicon Ultra Centrifugal Filters, 15 mL, 10K MWCO; pack of 24
Representative image only.
EMD Millipore Mfr # UFC901024 – Item # RK-29969-82
Low-binding regenerated cellulose membrane, fast processing, and >90% recoveries
  • Unique vertical housing
  • Designed with 15-mL volume and 10,000 Nominal molecular weight limit (NMWL)
  • 1 year warranty
PRODUCT OPTIONS
$548.10CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Tube Size (mL)15
  • Nominal Molecular Weight Limit (Mwl) (Dalton)10000
  • DescriptionAmicon Ultra Centrifugal Filters, 15 mL 10K 24PK
  • Warranty1 year
1 Year Warranty
Key Features
  • Unique vertical housing
  • Designed with 15-mL volume and 10,000 Nominal molecular weight limit (NMWL)
  • 1 year warranty
More About this Item
These units are premier tools for protein concentration and desalting. Typical applications include purifying macromolecular components from tissue culture extracts or cell lysates and concentrating biological samples.