EMD Millipore PLBC04310 Ultrafiltration Disc Membranes, 3k 43mm 10pk
Representative image only.
EMD Millipore Mfr # PLBC04310 – Item # RK-29949-12
Ideal for stirred cells and micropartition system
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
PRODUCT OPTIONS
$392.29CAD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (mm)44.5
  • Nominal Molecular Weight Limit (Mwl) (Dalton)3000
  • Diameter (cm)4.45
  • DescriptionUltrafiltration Disc Membranes, 3k 43mm 10pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
More About this Item
These high-recovery hydrophilic membranes are made of regenerated cellulose. Use when concentrating or desalting extremely dilute solutions or whenever your sample is hydrophobic.