EMD Millipore PLAC02510 Ultrafiltration Disc Membranes, 1k 25mm 10pk
Representative image only.
EMD Millipore Mfr # PLAC02510 – Item # RK-29949-00
Ideal for stirred cells and micropartition system
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
PRODUCT OPTIONS
$251.95CAD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (mm)25
  • Nominal Molecular Weight Limit (Mwl) (Dalton)1000
  • Diameter (cm)2.5
  • DescriptionUltrafiltration Disc Membranes, 1k 25mm 10pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
More About this Item
These high-recovery hydrophilic membranes are made of regenerated cellulose. Use when concentrating or desalting extremely dilute solutions or whenever your sample is hydrophobic.