This item is now discontinued. Please browse similar items
EMD Millipore 14432 Ultrafiltration Disc Membranes, 100K; 10/Pk
Representative image only.
EMD Millipore Mfr # 14432 – Item # RK-29949-36
Ideal for stirred cells and micropartition system
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Nominal Molecular Weight Limit (Mwl) (Dalton)100000
  • Diameter (mm)44.5
  • Diameter (cm)4.45
  • DescriptionUltrafiltration Disc Membranes, 100K; 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
More About this Item
These high-recovery hydrophilic membranes are made of regenerated cellulose. Use when concentrating or desalting extremely dilute solutions or whenever your sample is hydrophobic.