This item is now discontinued. Please browse similar items
EMD Millipore 10003-1 Water Test Strips, Copper, 10 to 300 ppm, 100/Pk
Representative image only.
EMD Millipore Mfr # 1.10003.0001 – Item # RK-99572-14
Use for quick spot checks to ensure water quality
  • Convenient, on-site water testing
SPECS & DESCRIPTION
Specifications & Description
  • TestCopper
  • Measurement Points10 to 300 ppm
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionWater Test Strips, Copper, 10 to 300 ppm, 100/Pk
Key Features
  • Convenient, on-site water testing
More About this Item
These new water test strips are perfect for field testing, process control, and analytical and research applications. Strips are easy to use: simply dip the strip into water sample to be tested, remove strip, and compare resulting color change to enclosed color chart. Tests take approximately 1 to 10 minutes. Strips are packaged 100/pk.