This item is now discontinued. Please browse similar items
Electro-Tech Systems 5471-115V Dehumidifier with 2-1/2 lb dessicant column, 115 VAC
Representative image only.
Electro-Tech Systems Mfr # 5471-115V – Item # RK-37700-51
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)115
  • DescriptionDehumidifier with 2-1/2 lb dessicant column, 115 VAC
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
All models use Drierite® desiccant.
Model 37700-51 include 2-½ lb desiccant column and high output pump in an ABS enclosure. Get humidity below 5% RH in sealed chambers up to 20 cu ft. Order replacement Drierite for all models.