Economical Hemacytometer with Cover Slips, Dual Chamber
Representative image only.
Mfr # CTL-HEMM-GLDR – Item # RK-79001-00
Specially designed for counting erythrocytes and leukocytes
  • Two special counting chamber glass coverslips
  • Light transmission between 29 and 32%
  • 1 year warranty
$360.98CAD / Each
Enhance Protection Today!
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Number of slides)2
  • Sectioning / Trimming Settings0.1 mm Depth
  • DescriptionEconomical Hemacytometer with Cover Slips, Dual Chamber
  • Warranty1 Year
1 Year Warranty
Key Features
  • Two special counting chamber glass coverslips
  • Light transmission between 29 and 32%
  • 1 year warranty
More About this Item
This hemacytometer features high light transmission and brightly illuminated line. Double rulings are visible through a wide range of illumination. Neubauer rulings are 0.1 mm below cover glass. The H-shaped moat offers two counting areas. Fused rhodium on glass provides distinct lines.