This item is now discontinued. Please browse similar items
Ebro 1340-5437 IP-67 RTD Thermometer with Cert of Conformance, Blunt Tip; -148 to 932F
Representative image only.
Ebro Mfr # 1340-5437 – Item # RK-90111-13
Ideal for use in harsh environments like labs, production and the food industry
  • IP67-rated housing and key pad
  • Compact size allows for convenient storage
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-148
  • Max Temperature (° F)932
  • Min Temperature (° C)-100
  • Max Temperature (° C)500
  • Accuracy±0.2°F; ±0.4°F above 211° and below -58°F [±0.1°C; ±0.2°C above 99.99° and below -50°C]
  • Resolution0.04°F; 0.4°F above 212°F [0.02°C; 0.2°C above 100°C]
  • Low Battery IndicatorYes
  • Display Type0.3\" Display, 4-digit LCD
  • Height (in)4 1/2
  • No. of Probe(s) Included1
  • Battery Life5 years
  • Battery3v Battery (1)
  • DescriptionIP-67 RTD Thermometer with Cert of Conformance, Blunt Tip; -148 to 932F
  • Warranty2 year
Two year warrantyConformite EuropeenneIP 67
Key Features
  • IP67-rated housing and key pad
  • Compact size allows for convenient storage
  • 2 year warranty
More About this Item
These meters feature a robust splash/impact-resistant ABS housing, corrosion-resistant, stainless steel probe, battery life indicator and an auto shutoff. A large LCD displays the temperature readings and is dishwasher resistant. Multifunction models have max/min and hold capabilities. Available with a choice of probes that are per installed; blunt-tip for general-purpose use, penetration for semi soft materials and a remote sensor with perpetrating end for increased flexibility