DescriptionLabelWriter 450, Uses One Label Writer Label
Warranty2 Years
Key Features
Direct thermal print technology eliminates ribbons, toner, and ink cartridges
Automatically checks and corrects U.S. addresses
2 year warranty
More About this Item
Print crystal-clear text, graphics, and barcodes with 600 x 300 dpi resolution graphics. Printers allow you to choose from 60+ label styles and layouts just by browsing all available layouts on screen – quickly find the best label for any job. Label printers eliminate the hassle of sheet-fed labels by printing one label at a time, a few labels, or many labels in as little as two seconds each. Label printers handle many different label styles to accommodate numerous job functions — shipping and return labels, file folder labels, name badges, inventory, and bar code labels — and come with their own easy-to-use software that integrates seamlessly with Microsoft® Word or Outlook®, QuickBooks®, CardScan®, and other programs, without retyping. LabelWriter allows you to verify U.S. addresses and ZIP+4® codes with included Address Fixer™ software (requires Internet connection).
LabelWriter 450 prints basic labels for envelopes, packages, files inventory, bar codes, and more at speeds up to 51 labels per minute. Accepts LW labels.
LabelWriter 450 Twin Turbo holds two label rolls at once for maximum efficiency – print 60+ label layouts at speeds up to 71 labels per minute, without switching label rolls. Accepts LW labels.
LabelWriter 450 Duo prints 60+ paper label styles at speeds up to 71 labels per minute, and uses LW labels and D1 cassette labels.
System requirements: Windows® XP/Vista (32-bit, 64-bit) or Mac® OS 10.4 or higher