This item is now discontinued. Please browse similar items
Dwyer CXA-S2 Water pump pressure switch, NC, range 30-100 psig (2.1-6.9 bar)
Representative image only.
Dwyer Mfr # CXA-S2 – Item # RK-68063-44
Pump control for water systems
  • Suitable for all types of electrically driven water pumps: jets, submersible, reciprocating
  • Direct acting (NC) or reverse acting (NO) available depending on switch type and model number
  • Simple to install - mount in any orientation
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5 psig (±0.3 bar)
  • Min Pressure (PSI)30
  • Max Pressure (PSI)100
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsSilicone, Stainless Steel, steel
  • Process Connection1/4\" NPT(F)
  • Electrical ConnectionsScrew terminal
  • Switch output20 A at 120 VAC, 12 A at 240 VAC, 9.6 A at 240 VAC (3-phase), 8.6 A at 32 VDC, 3.1 A 120 VDC, 1.6 A at 240 VDC, snap action (NC)
  • OutputSPST
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionWater pump pressure switch, NC, range 30-100 psig (2.1-6.9 bar)
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Suitable for all types of electrically driven water pumps: jets, submersible, reciprocating
  • Direct acting (NC) or reverse acting (NO) available depending on switch type and model number
  • Simple to install - mount in any orientation
  • 1 year warranty
More About this Item
Dwyer Series CXA water pump pressure switches have been proven reliable for controlling automatic water systems-ideal for water well pumps and pumping systems. Easily adjust set point and deadband via screws inside the cover.