This item is now discontinued. Please browse similar items
Dwyer A-240-A Cooling Tower for Process Gauges, perforated style, 1/4" NPT
Representative image only.
Dwyer Mfr # A-240-A – Item # RK-68332-01
Protect your pressure instruments from high temperatures
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Pressure (PSI)0
  • Max Pressure (PSI)5800
  • Wetted Materials316 SS
  • Process Connection1/4\" NPT(M/F)
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionCooling Tower for Process Gauges, perforated style, 1/4\" NPT
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These perforated and spiral cooling towers protect pressure instruments during high temperature applications - ideal for use with pressure gauges, switches, and transmitters where the process media temperature exceeds the rating of the instrument. Choose the perforated style when a protective shell surrounding the stainless steel tube. Choose the spiral style for applications where no protective shell is required. Both the spiral and perforated styles are made with 316L stainless steel.