This item is now discontinued. Please browse similar items
Dwyer 641-18 Transmitter 18" probe length, no display
Representative image only.
Dwyer Mfr # 641-18 – Item # RK-10778-78
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe length (in)18
  • DescriptionTransmitter 18\" probe length, no display
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This air velocity transmitter is the ideal instrument for monitoring air flow. This transmitter uses a heated mass flow sensor, which allows for precise velocity measurements at various flow rates and temperatures. The transmitter’s 16 field-selectable ranges allow the versatility to be selected for several airflow applications. The LED display version produces a complete, low-cost solution for local indication of airflow.
FEATURES
  • Ranges to 15,000 FPM or 75 MPS
  • Optional bright LED display
  • Easy push button set-up
  • Compact housing
  • 4-20mA output
  • Digital filter for signal damping