This item is now discontinued. Please browse similar items
Dwyer 541-0014 Temperature/Process Controller Inductive/Capacitive Snubber
Representative image only.
Dwyer Mfr # 541-0014 – Item # RK-89800-99
Compatible with the Love B- and C-Series Temperature/Process Controllers
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTemperature/Process Controller Inductive/Capacitive Snubber
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
The Love B- and C-Series Temperature/Process Controllers with output relay require snubber 89800-99 when connecting to an inductive load; order contoller separately.