This item is now discontinued. Please browse similar items
Dwyer 2-5005 Minihelic Differential Pressure Gauge, 0-5.0" WC
Representative image only.
Dwyer Mfr # 2-5005 – Item # RK-68062-18
Gauge provides excellent readability in a compact size
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% full-scale at 70°F (21°C)
  • Dial Size (in)3
  • Dial Size (mm)76
  • Min Pressure (in WC)0
  • Max Pressure (in WC)5
  • Min Temperature (° F)20
  • Max Temperature (° F)140
  • Min Temperature (° C)7
  • Max Temperature (° C)60
  • Wetted MaterialsSilicone rubber
  • CaseGlass-filled nylon
  • Lens MaterialPolycarbonate
  • Process Connection3/16\" hose barb
  • Unit Of Measurement\" WC
  • DisplayAnalog dial
  • Media CompatibilityAir and noncombustible, compatible gases
  • DescriptionMinihelic Differential Pressure Gauge, 0-5.0\" WC
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Combining clean design, small size, and low cost with enough accuracy for all but the most demanding applications, the Minihelic® II gauge offers the latest in design features for a dial type differential pressure gauge . It is a very compact gauge but is easy to read and can safely operate at total pressures up to 30 psig. The gauge is designed for panel mounting in a single 2-5/8' diameter hole. Standard pressure connections are barbed fittings for 3/16' I.D. tubing. Over-pressure protection is built into the gauge by means of a blow-out membrane molded in conjunction with the diaphragm. Accidental over-ranging up to the rated total pressure will not damage the gauge . With removable lens and rear housing, the gauge may be easily serviced at minimum cost. With the housing