This item is now discontinued. Please browse similar items
Dwyer 166-6 Stainless Steel Pitot Tube 6" Insertion Length 1/8" Diameter
Representative image only.
Dwyer Mfr # 166-6 – Item # RK-10778-88
Almost impossible to damage
  • Hemispherical tips—withstand almost anything
  • Precision-made to AMCA and ASHRAE codes—two diameters, eleven models
  • Air velocity calculator
  • Direct-reading velocity charts
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe length (in)6
  • Probe Diameter (in)1/8
  • DescriptionStainless Steel Pitot Tube 6\" Insertion Length 1/8\" Diameter
  • Warranty1 year
1 Year Warranty
Key Features
  • Hemispherical tips—withstand almost anything
  • Precision-made to AMCA and ASHRAE codes—two diameters, eleven models
  • Air velocity calculator
  • Direct-reading velocity charts
  • 1 year warranty
More About this Item
Recommended for ducts 10" and larger in diameter. Plainly inscribed inch graduations on both sides show depth of insertion at a glance—this helps you find the exact center of any duct and make an accurate traverse quickly. Tubes comply with AMCA and ASHRAE specifications. Test-confirmed unity coefficients. Life-time construction of 304 stainless steel. Suitable for use in temperatures up to 1500°F (815°C). Accurate readings when tip is directed within 15° of airflow. Static connection extends 31⁄2" to serve as guide for directing tip. Swaged tip, silver soldered connections.