This item is now discontinued. Please browse similar items
DWK Life Sciences (Kimble) Quartz Crucible Cover, fits 34526-50 and -52. Case of 1
Representative image only.
DWK Life Sciences (Kimble) Mfr # 319012-0050 – Item # RK-34526-54
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialQuartz
  • AutoclavableYes
  • Inside Rim Diameter (mm)49
  • DescriptionQuartz Crucible Cover, fits 34526-50 and -52. Case of 1
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Quartz crucible cover replacement fits crucibles 34526-50 and 34526-52. Quartz material satisfies high-temperature requirements up to 1922°F (1050°C) — perfect for muffle furnace ashing procedures and ensures chemical purity for subsequent analysis.